Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM Reag. Ph. Eur., super gradient grade for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

Filtered through a 0.2 µm filter, packaged under nitrogen.

BDH83639.400 BDH83639.100E BDH83639.402
BDH83639.400CS 1315.85 USD
BDH83639.400 BDH83639.100E BDH83639.402
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM Reag. Ph. Eur., super gradient grade for HPLC, VWR Chemicals BDH®
Acetonitrile

From the exclusive family of high-quality, competitively priced BDH Chemicals – High Purity solvents for HPLC applications.


  • Designed to meet demanding HPLC and UV/Vis analytical and quality control requirements
  • Filtered down to 0.2 micron level and bottled under nitrogen
  • Feature tamper-evident caps for added peace of mind
  • Carry Globally Harmonized System (GHS) labeling to help with compliance
  • Feature environmentally friendly corrugated/pulp case inserts that are fully recyclable


Ordering information: To purchase these solvents in stainless steel returnable drums as part of the VWR ReCAP program, please contact your local VWR sales representative. A separate contract for VWR ReCAP products is required.

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6
Melting Pt: -45.7
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.9 %
Solution (100 g/L) Neutral
Acidity Max. 0.0008 meq/g
Alkalinity Max. 0.0006 meq/g
Coloration Max. 10 APHA
n 20/D 1.343 to 1.345
Evaporation Residue Max. 2 ppm
Water Max. 30 ppm
Absorbance (200 nm) Max. 0.10
Transmittance (190 nm) Min. 10 %
Transmittance (195 nm) Min. 80 %
Transmittance (200 nm) Min. 95 %
Transmittance (210 nm) Min. 96 %
Transmittance (220 nm) Min. 97 %
Transmittance (230 nm) Min. 98 %
Transmittance (240 nm) Min. 99 %
Transmittance (250 nm) Min. 99 %
Transmittance (240 to 420 nm) Min. 99 %
Fluorescence (as quinine) (254 nm) Max. 1 ppb
Gradient grade (210 nm) Max. 10 mAU
Conforms to ACS Passes Test
Conforms to ACS (HPLC/UV) Passes Test
Conforms to Reag. Ph.Eur. Passes Test
Conforms to USP Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR