N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

BDH83636.400
BDH83636.400CS 1332.86 USD
BDH83636.400
N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
N,N-Dimethylacetamide
Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 166.1 °C (1013 hPa)
Melting Pt: –20 °C
Flash Pt: 70 °C
MDL Number: MFCD00008686
CAS Number: 127-19-5
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) Min 99.5%
Residue after evaporation Max 5 ppm
Water (by KF coulometry) Max 0.03%
Transmittance at 270 nm Min 10 T(%)
Transmittance at 280 nm Min 50 T(%)
Transmittance at 290 nm Min 70 T(%)
Transmittance at 310 nm Min 90 T(%)
Transmittance at 320 nm Min 93 T(%)
Transmittance at 360 nm Min 97 T(%)
Transmittance at 400 nm Min 97 T(%)
Optical absorbance at 270 nm Max 1.00 AU
Optical absorbance at 280 nm Max 0.30 AU
Optical absorbance at 290 nm Max 0.15 AU
Optical absorbance at 320 AU Max 0.05 AU
Optical absorbance at 310 nm Max 0.03 AU
Optical absorbance at 360 nm Max 0.01 AU
Optical absorbance at 400 nm Max 0.01 AU

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR