1-Propanol ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International

BDH83635.400
BDH83635.400CS 1899.78 USD
BDH83635.400
1-Propanol ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
1-Propanol
Formula: H₃CCH₂CH₂OH
Boiling Pt: 97 °C (1013 hPa)
Melting Pt: –127 °C
Flash Pt: 15 °C
MDL Number: MFCD00002941
CAS Number: 71-23-8
UN: 1274
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) Min 99.5%
Color Max 10 APHA
Residue after evaporation Max 5 ppm
Titrable acid Max 0.0004 meq/g
Water (by KF coulometry) Max 0.08%
Transmittance at 210 nm Min 10 T(%)
Transmittance at 225 nm Min 30 T(%)
Transmittance at 250 nm Min 90 T(%)
Transmittance at 300 nm Min 99 T(%)
Transmittance at 400 nm Min 99 T(%)
Optical absorbance at 210 nm Max 1.00 AU
Optical absorbance at 225 nm Max 0.50 AU
Optical absorbance at 250 nm Max 0.05 AU
Optical absorbance at 300 nm Max 0.005 AU
Optical absorbance at 400 nm Max 0.005 AU


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR