Cyclohexane ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH83629.400
BDH83629.400CS 1181.92 USD
BDH83629.400
Cyclohexane ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Cyclohexane
Formula: C₆H₁₂
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
Water (by KF coulometry) Max. 0.01 %
Nonvolatile Residue Max. 0.0005 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Transmittance (230 nm) Min. 75 %
Transmittance (240 nm) Min. 80 %
Transmittance (250 nm) Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR