1,4-Dioxane ≥99.9%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

BDH83628.400
BDH83628.400CS 2071.47 USD
BDH83628.400
1,4-Dioxane ≥99.9%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
1,4-Dioxane
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 101.1
Melting Pt: 11.8
Flash Pt: 12 °C
MDL Number: MFCD00006571
CAS Number: 123-91-1
UN: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) Min 99.9%
Color Max 10 APHA
Peroxides/Epoxides (as Hydrogen peroxide at time of packaging) Max 0.5%
Residue after evaporation Max 5 ppm
Water (by KF coulometry) Max 0.5%
Transmittance at 215 nm Min 10 T(%)
Transmittance at 225 nm Min 25 T(%)
Transmittance at 250 nm Min 55 T(%)
Transmittance at 295 nm Min 98 T(%)
Transmittance at 400 nm Min 98 T(%)
Optical absorbance at 215 nm Max 1.00 AU
Optical absorbance at 225 nm Max 0.60 AU
Optical absorbance at 250 nm Max 0.25 AU
Optical absorbance at 295 nm Max 0.01 AU
Optical absorbance at 400 nm Max 0.01 AU


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR