Chloroform ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Stabilised with amylene

BDH83626.400
BDH83626.400CS 1521.52 USD
BDH83626.400
Chloroform ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Chloroform

Stabilized with Amylene

Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) Min. 99.8 %
Water (by KF coulometry) Max. 0.05 %
Residue after evaporation Max. 0.001 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Transmittance (250 nm) Min. 50 %
Transmittance (260 nm) Min. 84 %
Transmittance (280 nm) Min. 95 %
Transmittance (300 nm) Min. 98 %
Conforms to BDH 15283 Passes Test
Stabilizer (as Amylenes) 100 - 200 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR