Diethyl ether ≥99.9% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International


Stabilization: Stabilised with ethanol

BDH83624.400
BDH83624.400CS 819.32 USD
BDH83624.400
Diethyl ether ≥99.9% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Diethyl ether
Formula: (CH₃CH₂)₂O
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Flash Pt: –40 °C
MDL Number: MFCD00011646
CAS Number: 60-29-7
UN: 1155
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC-FID) (excluding Ethanol) Min 99.9%
Color Max 10 APHA
Peroxides/Epoxides (as Hydrogen peroxide at time of packaging) Max 5 ppm
Residue after evaporation Max 5 ppm
Stabilizer (as Ethyl alcohol) 1.5 - 2.5%
Water (by KF coulometry) Max 0.3%
Transmittance at 215 nm Min 10 T(%)
Transmittance at 230 nm Min 50 T(%)
Transmittance at 254 nm Min 80 T(%)
Transmittance at 280 nm Min 98 T(%)
Transmittance at 300 nm Min 99 T(%)
Optical absorbance at 215 nm Max 1.00 AU
Optical absorbance at 230 nm Max 0.30 AU
Optical absorbance at 254 nm Max 0.08 AU
Optical absorbance at 280 nm Max 0.01 AU
Optical absorbance at 300 nm Max 0.005 AU


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR