Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Acetic acid ethyl ester, Acetic ether, Acetoxyethane, Acetidin, Ethyl acetic ester, Ethyl ethanoate, EtOAc, Vinegar naphtha, Acetic acid ethyl ester

Filtered through a 0.2 µm filter, packaged under nitrogen.

BDH83621.400 CABDH83621.400
BDH83621.400CS 838.5 USD
BDH83621.400 CABDH83621.400
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: -83
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.8 %
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation Residue Max. 5 ppm
Water Max. 0.1 %
Transmittance (260 nm) Min. 70 %
Transmittance (270 nm) Min. 90 %
Transmittance (300 nm) Min. 96 %
Conforms to BDH 15248 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR