Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

BDH7996-4
BDH7996-4CS 744.23 USD
BDH7996-4
Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide

Made with deionized water.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Assay 49.0 - 51.0 %
Sodium carbonate 0.3 % max.
Chloride (Cl) 0.01 % max.
Heavy metals (as Ag) by EPA-6010B 0.001 % max.
Iron (Fe) by EPA-6010B 5 ppm max.
Nickel (Ni) 0.001 % max.
Nitrogen compounds (as N) 5 ppm max.
Phosphate (PO4) 5 ppm max.
Potassium (K) 0.05 % max.
Appearance/color Clear colourless liquid
Sulfate (SO4) 0.005 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR