Potassium chloride 4.0 M saturated with silver chloride aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDH7965-2
BDH7965-2CS 883.12 USD
BDH7965-2
Potassium chloride 4.0 M saturated with silver chloride aqueous solution, VWR Chemicals BDH®
Potassium chloride

Made with deionized water and saturated with <0.01% AgCl.

Formula: KCl
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Shelf-life 1 year
Appearance/color Clear liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR