Citric acid 10% in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

BDH7949-1
BDH7949-1CS 904.47 USD
BDH7949-1
Citric acid 10% in aqueous solution, VWR Chemicals BDH®
Citric acid

Made with deionized water.

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Pale yellow clear liquid
Shelf-life 2 years
Assay (w/v) 9.50 - 10.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR