Trichloroacetic acid (TCA) 20% in aqueous solution analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

BDH7930-2
BDH7930-2CS 1255.31 USD
BDH7930-2
Trichloroacetic acid (TCA) 20% in aqueous solution analytical reagent, VWR Chemicals BDH®
Trichloroacetic acid (TCA)

Made with purified water.

Formula: Cl₃CCOOH
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: ≤1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 2564
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Prepared to formulation on file Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR