L(+)-Tartaric acid solution 10% (w/v), VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
BDH7923-2
BDH7923-2CS
912.71
USD
BDH7923-2
L(+)-Tartaric acid solution 10% (w/v), VWR Chemicals BDH®
L(+)-Tartaric acid
Formula:
C₄H₆O₆ MW: 150.09 g/mol |
MDL Number:
MFCD00064207 CAS Number: 87-69-4 |
Specification Test Results
Assay (w/v) | 9.70 - 10.30 % |
Appearance/color | Clear liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...