Sulfuric acid 2.5 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDH7922-1
BDH7922-1CS 829.3 USD
BDH7922-1
Sulfuric acid 2.5 N in aqueous solution, VWR Chemicals BDH®
Sulfuric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: H₂SO₄
MW: 98.08 g/mol
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 1
Normality 2.495 - 2.505 N
Shelf-life 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR