Silver nitrate 0.171 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

BDH7916-4
BDH7916-4CS 2706.09 USD
BDH7916-4
Silver nitrate 0.171 N in aqueous solution, VWR Chemicals BDH®
Silver nitrate

Made with deionized water.


Suitable as a titrant.

Formula: AgNO₃
MW: 169.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 919 X
Normality 0.1660 - 0.1760 N
Shelf-life 18 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR