Potassium hydroxide 10% (w/v) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

BDH7907-1
BDH7907-1CS 1001.98 USD
BDH7907-1
Potassium hydroxide 10% (w/v) in aqueous solution, VWR Chemicals BDH®
Potassium hydroxide

Made with deionized water.

Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 X
Shelf-life 2 years
Assay (w/v) 9.90 - 10.10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR