Magnesium chloride 51% (w/v) in aqueous solution, Reagent Grade analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

BDH7899-1
BDH7899-1CS 2461.81 USD
BDH7899-1
Magnesium chloride 51% (w/v) in aqueous solution, Reagent Grade analytical reagent, VWR Chemicals BDH®
Magnesium chloride

Made with deionized water.

Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: 1412
Melting Pt: 715
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Appearance/color Clear to slightly turbid liquid
Shelf-life 2 years
Magnesium chloride 48.5 - 53.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR