Ammonia solution 20% (v/v) (1+4), VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH7868-1
BDH7868-1CS 1945.03 USD
BDH7868-1
Ammonia solution 20% (v/v) (1+4), VWR Chemicals BDH®
Ammonia solution

Per liter formula:
200mLs ACS Ammonium Hydroxide 28-30% W/W
800mLs Deionized Water

Formula: NH₃
Density: 0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR