EDTA solution, 0.5M, pH 8.0
Supplier: VWR International
BDH7830-1
BDH7830-1CS 2651.8 USD
BDH7830-1
EDTA solution, 0.5M, pH 8.0
Buffers Cloning Buffers
N.I.S.T traceable to SRM 919.
Suitable as a titrant or testing standard.