Potassium thiocyanate 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Potassium rhodanide

BDH7807-1
BDH7807-1CS 2646.26 USD
BDH7807-1
Potassium thiocyanate 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Potassium thiocyanate

Made with deionized water.

Formula: KSCN
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1

Order Now

Specification Test Results

Normality 0.0995 - 0.1005 N
Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 919 Yes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR