Polysorbate 80 solution analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate

BDH7781-2
BDH7781-2CS 1681.21 USD
BDH7781-2
Polysorbate 80 solution analytical reagent, VWR Chemicals BDH®
Polysorbate 80
Melting Pt: 100
Flash Pt: 113
MDL Number: MFCD00082107
CAS Number: 9005-65-6

Order Now

Specification Test Results

Appearance/color Clear, yellow to dark yellow liquid
Identification Passes test
Acid value 2.0 max.
Hydroxyl value 65 - 80
Saponification value 45 - 55
Specific gravity (25°C/25 °C) 1.060 - 1.090
Viscosity (25 °C) 300 - 500 cSt
Heavy metals (as Pb) 10 ppm max.
Residue after ignition 0.25 % max.
Water 3.0% max.
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR