Tris-EDTA solution (TE solution), VWR Chemicals BDH®
Supplier: VWR International
BDH7732-1
BDH7732-1CS
1588.82
USD
BDH7732-1
Tris-EDTA solution (TE solution), VWR Chemicals BDH®
Tris-EDTA solution (TE solution)
Liquid. Contains ultra pure deionized water, 0.5M EDTA with pH 7.5, and 1M TRIS with pH 8.
Specification Test Results
Appearance/color | Clear liquid |