Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) solution 12,1% (w/v) 1 M analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

BDH7731-1
BDH7731-1CS 3012.01 USD
BDH7731-1
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) solution 12,1% (w/v) 1 M analytical reagent, VWR Chemicals BDH®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Made with deionized water ultra pure. pH 7.5.

Formula: NH₂C(CH₂OH)₃
MDL Number: MFCD00004679
CAS Number: 77-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Clear
Shelf-life 18 months
pH 7.45 - 7.55


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR