Potassium hydroxide 0.5 N in methanol, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

BDH7730-1
BDH7730-1CS 1343.31 USD
BDH7730-1
Potassium hydroxide 0.5 N in methanol, VWR Chemicals BDH®
Potassium hydroxide

Standardized at 25 °C against chemicals whose certification is traceable to NIST.

Formula: KOH
MW: 56.11 g/mol
Flash Pt: 11
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 X
Normality 0.495 - 0.505 N
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR