Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®

Supplier: VWR International

BDH7637-1
BDH7637-1CS 1153.44 USD
BDH7637-1
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Hydrogen peroxide

Made with deionized water.

Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: 100 °C (1013 hPa)
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 8040 1
Assay 3.00 - 3.50 %
Stabilizer (Acetanilide) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR