EDTA disodium salt dihydrate solution 0.25 M, Reagent Grade analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
BDH7590-2
BDH7590-2CS
3591.18
USD
BDH7590-2
EDTA disodium salt dihydrate solution 0.25 M, Reagent Grade analytical reagent, VWR Chemicals BDH®
EDTA disodium salt dihydrate
Suitable as a titrant. Made with deionized water.
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 |
|
Specification Test Results
Molarity | 0.2495 - 0.2505 M |
N.I.S.T. Traceable to SRM 915 | Yes |
Appearance/color | Clear liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...