Potassium hydroxide 0.1 N in ethanol, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

BDH7457-1
BDH7457-1CS 891.02 USD
BDH7457-1
Potassium hydroxide 0.1 N in ethanol, VWR Chemicals BDH®
Potassium hydroxide

Made with 200 proof ethyl alcohol. Prepared to ASTM D 94.

Formula: KOH
MW: 56.11 g/mol
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance/color Clear to slightly turbid liquid
Molarity 0.0995 - 0.1005 M
Shelf-life 1 year
Prepared conforms to ASTM D94 X

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR