Hydrochloric acid 3.0 N, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

BDH7375-1
BDH7375-1CS 745.32 USD
BDH7375-1
Hydrochloric acid 3.0 N, VWR Chemicals BDH®
Hydrochloric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 1
Normality 2.994 - 3.006 N
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR