Trichloroacetic acid (TCA) 95.0-105.0% (w/v) (6.12M), VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

BDH7372-2
BDH7372-2CS 2810.61 USD
BDH7372-2
Trichloroacetic acid (TCA) 95.0-105.0% (w/v) (6.12M), VWR Chemicals BDH®
Trichloroacetic acid (TCA)

Made with deionized water.

Formula: Cl₃CCOOH
MW: 163.39 g/mol
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
Merck Index: 13,09700
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 2 years
Trichloroacetic acid (w/v) 98 - 102 %
Molarity 5.998 - 6.242 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR