Silver nitrate solution 0.0141 N, VWR Chemicals BDH®

Supplier: VWR

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

BDHVBDH7359-1
BDH7359-1CS 2331.14 USD
BDH7359-1 BDH73591
Silver nitrate solution 0.0141 N, VWR Chemicals BDH®
Silver nitrate

Suitable as a titrant

Formula: AgNO₃
MW: 169.87 g/mol
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 919 X
Normality 0.0136 - 0.0146 N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR