Potassium chromate 5% (w/v) in aqueous solution analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

BDH7350-2
BDH7350-2CS 608.56 USD
BDH7350-2
Potassium chromate 5% (w/v) in aqueous solution analytical reagent, VWR Chemicals BDH®
Potassium chromate

For the titration of chlorine using silver nitrate.

Formula: K₂CrO₄
MW: 194.19 g/mol
Boiling Pt: 1000
Melting Pt: 985
MDL Number: MFCD00011368
CAS Number: 7789-00-6
EINECS: 232-140-5
UN: 3287
ADR: 6.1 III
Merck Index: 12,07784

Order Now

Specification Test Results

Free from chlorine Pass
For titration of chloride using silver nitrate Suitable
Appearance/color Clear yellow liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR