Potassium Chloride, Solution 0.01M, Conductivity Standard 1413 µmhos/cm, VWR Chemicals BDH®
Supplier: VWR International
BDH7349-1
BDH7349-1CS 835.5 USD
BDH7349-1
Potassium Chloride, Solution 0.01M, Conductivity Standard 1413 µmhos/cm, VWR Chemicals BDH®
Standards Conductivity Standards
Conductivity standard: 1413 µS/m.