Orthophosphoric acid 10% (v/v) (1+9), VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Phosphoric acid

BDH7346-1
BDH7346-1CS 708.17 USD
BDH7346-1
Orthophosphoric acid 10% (v/v) (1+9), VWR Chemicals BDH®
Orthophosphoric acid

Per liter formula:
100mLs ACS Phosphoric Acid minimum 85% W/W
900mLs Deionized Water

Formula: H₃PO₄
MW: 98 g/mol
Density: ∼1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 2 years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR