Sulfuric acid 25% (v/v) (1+3), VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH7339-1
BDH7339-1CS 797.69 USD
BDH7339-1
Sulfuric acid 25% (v/v) (1+3), VWR Chemicals BDH®
Sulfuric acid

Per liter formula:
250mLs ACS Sulfuric Acid 95-98% W/W
750mLs Deionized Water

Formula: H₂SO₄
MW: 98.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 2 years
Density 1.240 - 1.270 at 25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR