Oxalic acid 10% (w/v) in aqueous solution analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH7336-1
BDH7336-1CS 1655.75 USD
BDH7336-1
Oxalic acid 10% (w/v) in aqueous solution analytical reagent, VWR Chemicals BDH®
Oxalic acid

Made with deionized water.

Formula: C₂H₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
Assay (w/v) 9.75 - 10.25 %
Shelf-life 1 year


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR