Nitric acid 50% (v/v) (1+1), VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Nitric acid solution

BDH7335-1
BDH7335-1CS 707.89 USD
BDH7335-1
Nitric acid 50% (v/v) (1+1), VWR Chemicals BDH®
Nitric acid

Per liter formula:
500mLs ACS Nitric Acid 68-70% W/W
500mLs Deionized Water

Formula: HNO₃
MW: 63.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 18 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR