Iron(III) chloride solution 0,025% (w/v) analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Iron trichloride, Ferric chloride

BDH7318-1
BDH7318-1CS 797.52 USD
BDH7318-1
Iron(III) chloride solution 0,025% (w/v) analytical reagent, VWR Chemicals BDH®
Iron (III) chloride
Formula: FeCl₃
MDL Number: MFCD00011005
CAS Number: 7705-08-0
UN: 1773
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear orange liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR