Fehling's reagent I (copper(II) sulfate solution) 3.46% (w/v), VWR Chemicals BDH®

Supplier: VWR International

BDH7316-1
BDH7316-1CS 719.16 USD
BDH7316-1
Fehling's reagent I (copper(II) sulfate solution) 3.46% (w/v), VWR Chemicals BDH®
Fehling's reagent I (Copper (II) sulphate solution)

Made with deionized water.

Formula: CuSO₄
Boiling Pt: ? 100 °C (1013 hPa)
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Shelf-life 2 years
Appearance/color Clear blue liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR