Calcium chloride 2.75% (w/v) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDH7308-1
BDH7308-1CS 1547.92 USD
BDH7308-1
Calcium chloride 2.75% (w/v) in aqueous solution, VWR Chemicals BDH®
Calcium chloride

Made with deionized water.

Formula: CaCl₂
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 1 year
Assay (w/v) 2.73 - 2.77 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR