Potassium chloride 3.0 M in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDH7296-0
BDH7296-0CS 970.59 USD
BDH7296-0
Potassium chloride 3.0 M in aqueous solution, VWR Chemicals BDH®
Potassium chloride

Made with deionized water.

Formula: KCl
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR