Sodium chloride solution 0.90% (w/v) in water, VWR Chemicals BDH®

Supplier: VWR International

BDH7257-1 BDH7257-2 BDH7257-4 BDH7257-7
BDH7257-1CS 210.69 USD
BDH7257-1 BDH7257-2 BDH7257-4 BDH7257-7
Sodium chloride solution 0.90% (w/v) in water, VWR Chemicals BDH®
Sodium chloride

Made with deionized water

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3

Order Now

Specification Test Results

Assay (w/v) 0.89 - 0.91 %
Appearance Clear liquid
Traceable to NIST SRM 919 Confirmed
Prepared to formulation on file Confirmed
Shelf-life 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR