Cerium(IV) sulfate solution 0.1 N, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Ceric sulfate, Cerium disulfate, Cerium disulphate, Ceric sulphate, Cerium(IV) sulfate, Cerium(IV) sulphate

BDH7253-1
BDH7253-1CS 3521.03 USD
BDH7253-1 BDH7253-4
Cerium(IV) sulfate solution 0.1 N, VWR Chemicals BDH®
Cerium (IV) sulphate

Made with purified water and dilute ACS grade sulfuric acid.

Formula: Ce(SO₄)₂
MW: 332.24 g/mol
MDL Number: MFCD00148852
CAS Number: 13590-82-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Normality 0.0995 - 0.1005 N
Appearance/color Clear orange liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR