Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
BDH7246-1
BDH7246-4
BDH7246-7
BDH7246-8
BDH7246-9
BDH7246-1CS
785.53
USD
BDH7246-1
BDH7246-4
BDH7246-7
BDH7246-8
BDH7246-9
Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide
Clear liquid. Corrosive and toxic.
Formula:
NaOH MW: 40 g/mol Boiling Pt: 119…135 °C (1013 hPa) Melting Pt: 9…10 °C Density: 1.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance | Clear colourless liquid |
Assay | 49.0 - 51.0 % |
Cl (Chloride) | ≤ 0.01 % |
Fe (Iron) | ≤ 5 ppm |
Heavy Metals (as Ag) | ≤ 0.001 % |
K (Potassium) | ≤ 0.05 % |
Ni (Nickel) | ≤ 0.001 % |
Nitrogen Compounds (as N) | ≤ 5 ppm |
PO₄ (Phosphate) | ≤ 5 ppm |
SO₄ (Sulphate) | ≤ 0.005 % |
Sodium Carbonate | ≤ 0.3 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...