Hydrochloric acid ( 0.027 mol/l; 0.027 N) 0.01 N analytical reagent, VWR Chemicals BDH®

Supplier: VWR International

BDH7237-1
BDH7237-1CS 612.17 USD
BDH7237-1
Hydrochloric acid ( 0.027 mol/l; 0.027 N) 0.01 N analytical reagent, VWR Chemicals BDH®
Hydrochloric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: HCl
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable to SRM 723 Yes
Normality 0.0095 - 0.0105 N
Shelf-life 1 year


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR