Acetic acid solution 1.000 N ±0.002 analytical reagent

Supplier: VWR International

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

BDH7236-1
BDH7236-1CS 1433.81 USD
BDH7236-1
Acetic acid solution 1.000 N ±0.002 analytical reagent
Acetic acid

Standardized at 25 °C against chemicals whose certification is traceable to NIST. Made with high purity acid and deionized water. Suitable as a titrant.

Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 Yes
Normality 0.998 - 1.002 N
Shelf-life 1 year


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR