Sodium thiosulfate 0.10 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
BDH7228-4
BDH7228-7
BDH7228-1
BDH7228-4CS
400.56
USD
BDH7228-4
BDH7228-7
BDH72281
Sodium thiosulfate 0.10 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Sodium thiosulphate
Suitable as a titrant. Made with deionized water.
Formula:
Na₂S₂O₃ Density: 1.008 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003499 CAS Number: 7772-98-7 EINECS: 231-867-5 |
Specification Test Results
Appearance/color | Clear liquid |
N.I.S.T. traceable to SRM 136 | Confirmed |
Normality | 0.0995 - 0.1005 N |
Prepared to formulation on file | Confirmed |
— | — |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...