Sodium thiosulfate 0.025 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
BDH7227-1
BDH7227-4
BDH7227-1CS
681.6
USD
BDH7227-1
BDH7227-4
Sodium thiosulfate 0.025 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Sodium thiosulphate
Suitable as a titrant. Made with deionized water.
Formula:
Na₂S₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00003499 CAS Number: 7772-98-7 EINECS: 231-867-5 |
Specification Test Results
Appearance/color | Clear liquid |
N.I.S.T. traceable to SRM 136 | Yes |
Normality | 0.0245 - 0.0255 N |
Shelf-life | 6 Months |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...