Sodium hydroxide (1.3 - 12 mol/l; 1.3 - 12 N) in aqueous solution 5.0 N in aqueous solution analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

BDH7225-1 BDH7225-4 BDH7225-10
BDH7225-1CS 731.61 USD
BDH7225-1 BDH7225-4 BDH7225-10
Sodium hydroxide (1.3 - 12 mol/l; 1.3 - 12 N) in aqueous solution 5.0 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Sodium hydroxide

Made with deionized water. Suitable as a titrant.

Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 Yes
Normality 4.95 - 5.05 N
Shelf-life 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR