Sodium hydroxide 2.5 N in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

BDH7224-1
BDH7224-1CS 1301.54 USD
BDH7224-1
Sodium hydroxide 2.5 N in aqueous solution, VWR Chemicals BDH®
Sodium hydroxide

AOAC and APHA. Suitable as a titrant.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. traceable to SRM 84 1
Normality 2.40 - 2.60 N
Shelf-life 1 year

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR