Sodium hydroxide (0.13 - 0.55 mol/l; 0.13 - 0.55 N) in aqueous solution 0.2 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier:
VWR International
|
Warning
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
BDH7220-1
BDH7220-4
BDH7220-1CS
641.97
USD
BDH7220-1
BDH7220-4
Sodium hydroxide (0.13 - 0.55 mol/l; 0.13 - 0.55 N) in aqueous solution 0.2 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Sodium hydroxide
AOAC, APHA, and ASTM. Suitable as titrant.
Formula:
NaOH Boiling Pt: ∼105 °C (1013 hPa) Density: 1.02 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,III |
Specification Test Results
Appearance/color | Clear liquid |
N.I.S.T. traceable to SRM 84 | Yes |
Normality | 0.1995 - 0.2005 N |
Shelf-life | 18 months |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...